Läs mer
(S)-Metalaxyl, TRC
Produktkod. 30136380
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136380 500 mg
17963035 250 mg
17914735 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136380 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM258970500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 69516-34-3
Molekylformel C15 H21 N O4
Molekylvikt (g/mol) 279.332
Synonym N-(2,6-Dimethylphenyl)-N-(Methoxyacetyl)-L-Alanine Methyl Ester
IUPAC-namn methyl (2S)-2-(N-(2-methoxyacetyl)-2,6-dimethylanilino)propanoate
LEDER COCC(=O)N([C@@H](C)C(=O)OC)c1c(C)cccc1C
Specifikationer
Kemiskt namn eller material (S)-Metalaxyl
Kvantitet 500 mg
InChI formel InChI=1S/C15H21NO4/c1-10-7-6-8-11(2)14(10)16(13(17)9-19-4)12(3)15(18)20-5/h6-8,12H,9H2,1-5H3/t12-/m0/s1
Formel vikt 279.147