Läs mer
[S-Methanethiosulfonylcysteaminyl]ethylenediamine-N,N,N’,N’-Tetraacetic Acid(mixture of mono-MTS and bis-MTS) (~90%), TRC
Produktkod. 30468290
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30468290 10 mg
1 options
Produktkod. 30468290 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM25883510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material [S-Methanethiosulfonylcysteaminyl]ethylenediamine-N,N,N',N'-Tetraacetic Acid
CAS 832733-28-5
Molekylformel C13H23N3O9S2
Kvantitet 10 mg
Synonym 9,12-Bis(carboxymethyl)-7-oxo-2,3-dithia-6,9,12-triazatetradecan-14-oic acid 2,2-Dioxide, MTS-EDTA
InChI formel InChI=1S/C13H23N3O9S2/c1-27(24,25)26-5-2-14-10(17)6-15(7-11(18)19)3-4-16(8-12(20)21)9-13(22)23/h2-9H2,1H3,(H,14,17)(H,18,19)(H,20,21)(H,22,23)
LEDER O=C(CN(CCN(CC(NCCSS(=O)(C)=O)=O)CC(O)=O)CC(O)=O)O
IUPAC-namn 2,2'-((2-((carboxymethyl)(2-((2-((methylsulfonyl)thio)ethyl)amino)-2-oxoethyl)amino)ethyl)azanediyl)diacetic acid
Molekylvikt (g/mol) 429.46
Formel vikt 429.09