Läs mer
(S)-(+)-Methoprene, TRC
Produktkod. 17991815
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991815 1 g
17943815 100 mg
17901925 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17991815 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM2258151G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65733-16-6
Molekylformel C19 H34 O3
Molekylvikt (g/mol) 310.47
Synonym (S)-Methoprene, 2,4-Dodecadienoic acid, 11-methoxy-3,7,11-trimethyl-, 1-methylethyl ester, (2E,4E,7S)-, 2,4-Dodecadienoic acid, 11-methoxy-3,7,11-trimethyl-, 1-methylethyl ester, [S-(E,E)]-, (+)-Methoprene, (7S)-Methoprene, Altosid XR Briquets, Altoside CP 10, Altoside CP 2, Diacon II, Extinguish, Precor, S-Methoprene, ZR 2458, d-Methoprene
IUPAC-namn propan-2-yl (2E,4E,7S)-11-methoxy-3,7,11-trimethyldodeca-2,4-dienoate
LEDER COC(C)(C)CCC[C@H](C)C\C=C\C(=C\C(=O)OC(C)C)\C
Specifikationer
Kemiskt namn eller material (S)-Methoprene
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C19H34O3/c1-15(2)22-18(20)14-17(4)11-8-10-16(3)12-9-13-19(5,6)21-7/h8,11,14-16H,9-10,12-13H2,1-7H3/b11-8+,17-14+/t16-/m1/s1
Formel vikt 310.2508
Procent renhet >95%