Läs mer
(S)-Methyl 2-methylpyrrolidine-2-carboxylate hydrochloride, TRC
Produktkod. 30473535
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30473535 50 mg
1 options
Produktkod. 30473535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33861350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Methyl 2-methylpyrrolidine-2-carboxylate hydrochloride
CAS 220060-08-2
Molekylformel C7H13NO2 . HCl
Kvantitet 50 mg
Synonym alpha-Methyl L-Proline Methyl Ester Hydrochloride
InChI formel InChI=1S/C7H13NO2.ClH/c1-7(6(9)10-2)4-3-5-8-7;/h8H,3-5H2,1-2H3;1H/t7-;/m0./s1
LEDER Cl.COC(=O)[C@]1(C)CCCN1
IUPAC-namn methyl (2S)-2-methylpyrrolidine-2-carboxylate;hydrochloride
Molekylvikt (g/mol) 179.64
Formel vikt 179.0713