Läs mer
(S)-Methyl 3-(4,4-difluorocyclohexanecarboxamido)-3-phenylpropanoate, TRC
Produktkod. 30477096
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30477096 250 mg
1 options
Produktkod. 30477096 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM303771250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Methyl 3-(4,4-difluorocyclohexanecarboxamido)-3-phenylpropanoate
CAS 948592-19-6
Molekylformel C17H21F2NO3
Kvantitet 250 mg
Synonym Benzenepropanoic acid','β-[[(4,4-difluorocyclohexyl)carbonyl]amino]-','methyl ester','(βS)- (ACI), Methyl (βS)-β-[[(4,4-difluorocyclohexyl)carbonyl]amino]benzenepropanoate (ACI), (S)-Methyl 3-(4,4-Difluorocyclohexanecarboxamido)-3-Phenylpropanoate
InChI formel InChI=1S/C17H21F2NO3/c1-23-15(21)11-14(12-5-3-2-4-6-12)20-16(22)13-7-9-17(18,19)10-8-13/h2-6,13-14H,7-11H2,1H3,(H,20,22)/t14-/m0/s1
LEDER COC(=O)C[C@H](NC(=O)C1CCC(F)(F)CC1)c2ccccc2
IUPAC-namn methyl (3S)-3-[(4,4-difluorocyclohexanecarbonyl)amino]-3-phenylpropanoate
Molekylvikt (g/mol) 325.35
Formel vikt 325.1489