Läs mer
S-Methyl-isothiouronium-13C Hemisulfate, TRC
Produktkod. 30465867
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30465867 50 mg
1 options
Produktkod. 30465867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM31496350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-Methyl-isothiouronium-13C Hemisulfate
CAS 113899-62-0
Molekylformel C13CH6N2S . 1/2H 2SO4
Kvantitet 50 mg
Synonym Carbamimidothioic-13C Acid Methyl Ester Sulfate, S-Methyl-isothiouronium-13C Sulfate
InChI formel InChI=1S/C2H6N2S.H2O4S/c1-5-2(3)4;1-5(2,3)4/h1H3,(H3,3,4);(H2,1,2,3,4)/i2+1;
LEDER N[13C](SC)=N.O=S(O)(O)=O
Molekylvikt (g/mol) 140.18
Formel vikt 140.015