Läs mer
S-Methyl-L-thiocitrulline Dihydrochloride, TRC
Produktkod. 30478499
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30478499 10 mg
1 options
Produktkod. 30478499 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM33075010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-Methyl-L-thiocitrulline, Hydrochloride
CAS 156719-39-0
Molekylformel C7 H15 N3 O2 S . Cl H
Kvantitet 10 mg
InChI formel InChI=1S/C7H15N3O2S.ClH/c1-13-7(9)10-4-2-3-5(8)6(11)12;/h5H,2-4,8H2,1H3,(H2,9,10)(H,11,12);1H/t5-;/m1./s1
LEDER Cl.CSC(=N)NCCC[C@@H](N)C(=O)O
IUPAC-namn (2R)-2-amino-5-[(C-methylsulfanylcarbonimidoyl)amino]pentanoic acid;hydrochloride
Molekylvikt (g/mol) 241.74
Formel vikt 241.0652