Learn More
(S)-(-)-α-Methylbenzyl isocyanate, 98%, Thermo Scientific Chemicals
Catalog Number p-159273
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
14649-03-7
C9H9NO
147.18
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
14649-03-7
C9H9NO
147.18
MFCD00002036
JJSCUXAFAJEQGB-QMMMGPOBSA-N
s---1-phenylethyl isocyanate, 1s-1-isocyanatoethyl benzene, s---alpha-methylbenzyl isocyanate, benzene, 1s-1-isocyanatoethyl, isocyanic acid s---a-methylbenzyl ester, s-alpha-methylbenzyl isocyanate, s---1-phenylethylisocyanate, isocyanic acid s---alpha-methylbenzyl ester, isocyanic acid s---methylbenzyl ester, 1-isocyanatoethyl benzene #
2724173
[(1S)-1-isocyanatoethyl]benzene
CC(C1=CC=CC=C1)N=C=O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chiral derivatization reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0500g/mL
Boiling Point 55.0°C to 56.0°C (2.5 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH(CH3)NCO
Refractive Index 1.5130 to 1.5150
Quantity 1 mL
Specific Gravity 1.05
Specific Rotation Condition −10° (20°C neat)
Specific Rotation − 10.00
Solubility Information Solubility in water: decomposes
Formula Weight 147.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-α-Methylbenzyl isocyanate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-a-Methylbenzyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.