missing translation for 'onlineSavingsMsg'
Learn More

S-Metolachlor Metabolite CGA 50720, TRC

Product Code. 30483503
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30483503 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30483503 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM338708100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material S-Metolachlor CGA 50720
CAS 152019-74-4
Molekylformel C11 H13 N O3
Kvantitet 100 mg
Synonym S-Metolachlor CGA 50720, Acetic acid, [(2-ethyl-6-methylphenyl)amino]oxo- (9CI), 2-[(2-Ethyl-6-methylphenyl)amino]-2-oxoacetic acid, CGA 50720
InChI formel InChI=1S/C11H13NO3/c1-3-8-6-4-5-7(2)9(8)12-10(13)11(14)15/h4-6H,3H2,1-2H3,(H,12,13)(H,14,15)
LEDER CCc1cccc(C)c1NC(=O)C(=O)O
IUPAC-namn 2-(2-ethyl-6-methylanilino)-2-oxoacetic acid
Molekylvikt (g/mol) 207.23
Formel vikt 207.0895
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.