Läs mer
(S)-Mirabegron, TRC
Produktkod. 17982685
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982685 100 mg
17910885 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982685 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM364895100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1796931-48-0
Molekylformel C21 H24 N4 O2 S
Molekylvikt (g/mol) 396.5059
Synonym (S)-2-(2-Aminothiazol-4-yl)-N-(4-(2-((2-hydroxy-2-phenylethyl)amino)ethyl)phenyl)acetamide, Mirabegron Impurity 1, ent-Mirabegron, 4-Thiazoleacetamide, 2-amino-N-[4-[2-[[(2S)-2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]-, 2-Amino-N-[4-[2-[[(2S)-2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]-4-thiazoleacetamide, 2-(2-Amino-1,3-thiazol-4-yl)-N-[4-[2-[((2S)-2-hydroxy-2-phenylethyl)amino]ethyl]phenyl]acetamide
IUPAC-namn 2-(2-amino-1,3-thiazol-4-yl)-N-[4-[2-[[(2S)-2-hydroxy-2-phenylethyl]amino]ethyl]phenyl]acetamide
LEDER Nc1nc(CC(=O)Nc2ccc(CCNC[C@@H](O)c3ccccc3)cc2)cs1
Specifikationer
Kemiskt namn eller material (S)-Mirabegron
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C21H24N4O2S/c22-21-25-18(14-28-21)12-20(27)24-17-8-6-15(7-9-17)10-11-23-13-19(26)16-4-2-1-3-5-16/h1-9,14,19,23,26H,10-13H2,(H2,22,25)(H,24,27)/t19-/m1/s1
Formel vikt 396.162
Procent renhet >95%