Learn More
(S)-N-Benzyl-1,4-oxazepan-6-ol, TRC
Product Code. 30337338
Change view
Click to view available options
Kvantitet:
50 mg
Product Code. Kvantitet
30337338 50 mg
1 options
Product Code. 30337338 Supplier Toronto Research Chemicals Supplier No. TRCB28533550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material (S)-N-Benzyl-1,4-oxazepan-6-ol
CAS 943443-05-8
Molekylformel C12H17NO2
Kvantitet 50 mg
Synonym (6S)-Hexahydro-4-(phenylmethyl)-1,4-oxazepin-6-ol
InChI formel InChI=1S/C12H17NO2/c14-12-9-13(6-7-15-10-12)8-11-4-2-1-3-5-11/h1-5,12,14H,6-10H2/t12-/m0/s1
LEDER O[C@@H]1COCCN(Cc2ccccc2)C1
IUPAC-namn (6S)-4-benzyl-1,4-oxazepan-6-ol
Molekylvikt (g/mol) 207.27
Formel vikt 207.1259