Läs mer
(S)-N-(Benzyloxycarbonyl)-N-methylvaline, TRC
Produktkod. 30343393
Change view
Kvantitet:
5 g
Produktkod. Kvantitet
30343393 5 g
1 options
Produktkod. 30343393 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB2866305G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-N-(Benzyloxycarbonyl)-N-methylvaline
CAS 42417-65-2
Molekylformel C14H19NO4
Kvantitet 5 g
Synonym N-(Benzyloxycarbonyl)-N-methyl-L-valine, N-(Benzyloxycarbonyl)-N-methylvaline
InChI formel InChI=1S/C14H19NO4/c1-10(2)12(13(16)17)15(3)14(18)19-9-11-7-5-4-6-8-11/h4-8,10,12H,9H2,1-3H3,(H,16,17)/t12-/m0/s1
LEDER CC(C)[C@H](N(C)C(=O)OCc1ccccc1)C(=O)O
IUPAC-namn (2S)-3-methyl-2-[methyl(phenylmethoxycarbonyl)amino]butanoic acid
Molekylvikt (g/mol) 265.3
Formel vikt 265.1314