Läs mer
S-(+)-N-Desmethyl Mephenytoin, TRC
Produktkod. 30366652
Change view
Kvantitet:
5 mg
25 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366652 25 mg
30360711 250 mg
30360638 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30366652 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29207025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 65567-34-2
Molekylformel C11 H12 N2 O2
Molekylvikt (g/mol) 204.23
Synonym 2,4-Imidazolidinedione, 5-ethyl-5-phenyl-, (5S)-, 2,4-Imidazolidinedione, 5-ethyl-5-phenyl-, (S)-, (5S)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, (+)-5-Ethyl-5-phenylhydantoin, (+)-5-Phenyl-5-ethylhydantoin, (+)-Nirvanol, (S)-(+)-Nirvanol, (S)-5-Ethyl-5-phenylhydantoin, (S)-5-Ethyl-5-phenyl-2,4-imidazolidinedione, (S)-5-Phenyl-5-ethylhydantoin, (S)-Nirvanol
IUPAC-namn (5S)-5-ethyl-5-phenylimidazolidine-2,4-dione
LEDER CC[C@]1(NC(=O)NC1=O)c2ccccc2
Specifikationer
Kemiskt namn eller material S-(+)-N-Desmethyl Mephenytoin
Kvantitet 25 mg
InChI formel InChI=1S/C11H12N2O2/c1-2-11(8-6-4-3-5-7-8)9(14)12-10(15)13-11/h3-7H,2H2,1H3,(H2,12,13,14,15)/t11-/m0/s1
Formel vikt 204.0899