Läs mer
(S)-N-Methyl Pregabalin, TRC
Produktkod. 30481695
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30481695 25 mg
1 options
Produktkod. 30481695 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM32601525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-N-Methyl Pregabalin
CAS 1155843-61-0
Molekylformel C9H19NO2
Kvantitet 25 mg
Synonym Hexanoic acid, 5-methyl-3-[(methylamino)methyl]-, (3S)-, (3S)-5-Methyl-3-[(methylamino)methyl]hexanoic acid, (3S)-5-Methyl-3-[(methylamino)methyl]hexanoic Acid
InChI formel InChI=1S/C9H19NO2/c1-7(2)4-8(6-10-3)5-9(11)12/h7-8,10H,4-6H2,1-3H3,(H,11,12)/t8-/m0/s1
LEDER CNC[C@@H](CC(C)C)CC(=O)O
IUPAC-namn (3S)-5-methyl-3-(methylaminomethyl)hexanoic acid
Molekylvikt (g/mol) 173.25
Formel vikt 173.1416