Läs mer
(S)-N-Nitroso Anabasine (>98% ee), TRC
Produktkod. 30040630
Change view
Kvantitet:
1 mg
5 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040630 5 mg
30040382 1 mg
30007876 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040630 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN5242465MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1133-64-8
Molekylformel C10 H13 N3 O
Molekylvikt (g/mol) 191.23
Synonym (S)-N-Nitrosoanabasine, NAB, (S)-N-Nitroso Anabasine, Pyridine, 3-[(2S)-1-nitroso-2-piperidinyl]-, Anabasine, 1-nitroso- (8CI), Pyridine, 3-(1-nitroso-2-piperidinyl)-, (S)-, 3-[(2S)-1-Nitroso-2-piperidinyl]pyridine, (S)-N'-Nitrosoanabasine, 1-Nitrosoanabasine, N-Nitrosoanabasine, N'-Nitrosoanabasine
IUPAC-namn 3-[(2S)-1-nitrosopiperidin-2-yl]pyridine
LEDER O=NN1CCCC[C@H]1c2cccnc2
Specifikationer
Kemiskt namn eller material (S)-N-Nitrosoanabasine (NAB)
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C10H13N3O/c14-12-13-7-2-1-5-10(13)9-4-3-6-11-8-9/h3-4,6,8,10H,1-2,5,7H2/t10-/m0/s1
Formel vikt 191.1059
Procent renhet >95%