Läs mer
(S)-Naproxen Acyl-Beta-D-glucuronide, TRC
Produktkod. 30008063
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008063 5 mg
30006116 10 mg
30003421 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30008063 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN3775305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 41945-43-1
Molekylformel C20 H22 O9
Molekylvikt (g/mol) 406.38
Synonym 1-[(αS)-6-Methoxy-α-methyl-2-naphthaleneacetate] β-D-Glucopyranuronic Acid, (S)-Naproxen Glucuronide, Naproxen Glucuronide
IUPAC-namn (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-[(2S)-2-(6-methoxynaphthalen-2-yl)propanoyl]oxyoxane-2-carboxylic acid
LEDER COc1ccc2cc(ccc2c1)[C@H](C)C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O
Specifikationer
Kemiskt namn eller material Naproxen Acyl-beta-D-glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C20H22O9/c1-9(10-3-4-12-8-13(27-2)6-5-11(12)7-10)19(26)29-20-16(23)14(21)15(22)17(28-20)18(24)25/h3-9,14-17,20-23H,1-2H3,(H,24,25)/t9-,14-,15-,16+,17-,20-/m0/s1
Formel vikt 406.1264
Procent renhet >95%