missing translation for 'onlineSavingsMsg'
Läs mer

S-(-)-Nicotine Di-p-Toluoyl-D-Tartrate Salt, TRC

Produktkod. 30477020
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477020 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30477020 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN422001100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (-)-Nicotine Di-p-Toluoyl-D-Tartrate Salt
CAS 68935-26-2
Molekylformel C20 H18 O8 . C10 H14 N2
Kvantitet 100 mg
Synonym (2R,3R)-2,3-Bis[(4-methylbenzoyl)oxy]butanedioic acid compd. with 3-[(2S)-1-methyl-2-pyrrolidinyl]pyridine (1:1), 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioate (1:1), S-(-)-Nicotine Di-p-Toluoyl-L-Tartrate Salt
InChI formel InChI=1S/C20H18O8.C10H14N2/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14;1-12-7-3-5-10(12)9-4-2-6-11-8-9/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24);2,4,6,8,10H,3,5,7H2,1H3/t15-,16-;10-/m10/s1
LEDER CN1CCC[C@H]1c2cccnc2.Cc3ccc(cc3)C(=O)O[C@H]([C@@H](OC(=O)c4ccc(C)cc4)C(=O)O)C(=O)O
IUPAC-namn (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molekylvikt (g/mol) 548.58
Formel vikt 548.2159
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.