Läs mer
S-(-)-Nicotine Di-p-Toluoyl-D-Tartrate Salt, TRC
Produktkod. 30467999
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30467999 1 g
1 options
Produktkod. 30467999 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN4220011G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Nicotine Di-p-Toluoyl-D-Tartrate Salt
CAS 68935-26-2
Molekylformel C20 H18 O8 . C10 H14 N2
Kvantitet 1 g
Synonym (2R,3R)-2,3-Bis[(4-methylbenzoyl)oxy]butanedioic acid compd. with 3-[(2S)-1-methyl-2-pyrrolidinyl]pyridine (1:1), 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioate (1:1), S-(-)-Nicotine Di-p-Toluoyl-L-Tartrate Salt
InChI formel InChI=1S/C20H18O8.C10H14N2/c1-11-3-7-13(8-4-11)19(25)27-15(17(21)22)16(18(23)24)28-20(26)14-9-5-12(2)6-10-14;1-12-7-3-5-10(12)9-4-2-6-11-8-9/h3-10,15-16H,1-2H3,(H,21,22)(H,23,24);2,4,6,8,10H,3,5,7H2,1H3/t15-,16-;10-/m10/s1
LEDER CN1CCC[C@H]1c2cccnc2.Cc3ccc(cc3)C(=O)O[C@H]([C@@H](OC(=O)c4ccc(C)cc4)C(=O)O)C(=O)O
IUPAC-namn (2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molekylvikt (g/mol) 548.58
Formel vikt 548.2159