Läs mer
(S)-(-)-Nicotine Mono Tartrate, TRC
Produktkod. 30478691
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30478691 50 mg
1 options
Produktkod. 30478691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN41245550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (-)-Nicotine Mono Tartrate
CAS 6550-19-2
Molekylformel C10 H14 N2 . C4 H6 O6
Kvantitet 50 mg
Synonym 3-[(2S)-1-Methyl-2-pyrrolidinyl]pyridine (2R,3R)-2,3-dihydroxybutanedioate (1:1), Nicotine tartrate (1:1), (S)-3-(1-Methyl-2-pyrrolidinyl)pyridine [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1), (-)-Nicotine bitartrate, Nicotine d-bitartrate, Nicotine hydrotartrate, Nicotine tartrate, S-(-)-Nicotine D-Monotartrate
InChI formel InChI=1S/C10H14N2.C4H6O6/c1-12-7-3-5-10(12)9-4-2-6-11-8-9;5-1(3(7)8)2(6)4(9)10/h2,4,6,8,10H,3,5,7H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t10-;1-,2-/m01/s1
LEDER CN1CCC[C@H]1c2cccnc2.O[C@H]([C@@H](O)C(=O)O)C(=O)O
IUPAC-namn (2R,3R)-2,3-dihydroxybutanedioic acid;3-[(2S)-1-methylpyrrolidin-2-yl]pyridine
Molekylvikt (g/mol) 312.32
Formel vikt 312.1321