Läs mer
S-Nitroso-N-acetyl-D,L-penicillamine, TRC
Produktkod. 30477129
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30477129 25 mg
30474871 50 mg
30476505 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30477129 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN52250025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67776-06-1
Molekylformel C7 H12 N2 O4 S
Molekylvikt (g/mol) 220.25
Synonym Valine, N-acetyl-3-(nitrosothio)- (9CI, ACI), DL-Valine, N-acetyl-3-(nitrosothio)- (ZCI), N-Acetyl-3-(nitrosothio)valine (ACI), 2-Acetamido-3-methyl-3-(nitrosothio)butanoic acid, N-Acetyl-S-nitroso-DL-penicillamine, N-Acetyl-S-nitrosopenicillamine, S-Nitroso-N-acetyl-DL-penicillamine, S-Nitrosoacetylpenicillamine, SNAP, SNAP (amino acid), 2-Acetamido-3-methyl-3-nitrososulfanyl-butanoic acid, N-Acetyl-3-(nitrosothio)-DL-valine
IUPAC-namn 2-acetamido-3-methyl-3-nitrososulfanylbutanoic acid
LEDER CC(=O)NC(C(=O)O)C(C)(C)SN=O
Specifikationer
Kemiskt namn eller material S-Nitroso-N-acetyl-D,L-penicillamine
Kvantitet 25 mg
InChI formel InChI=1S/C7H12N2O4S/c1-4(10)8-5(6(11)12)7(2,3)14-9-13/h5H,1-3H3,(H,8,10)(H,11,12)
Formel vikt 220.0518