Läs mer
(S)-(-)-Nor Verapamil Hydrochloride, TRC
Produktkod. 30420383
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30420383 1 mg
1 options
Produktkod. 30420383 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN8774911MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-(-)-Norverapamil Hydrochloride
CAS 147158-72-3
Molekylformel C26 H36 N2 O4 . Cl H
Kvantitet 1 mg
Synonym Benzeneacetonitrile','α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-','monohydrochloride','(αS)- (9CI), Benzeneacetonitrile','α-[3-[[2-(3,4-dimethoxyphenyl)ethyl]amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)-','monohydrochloride','(S)-, (S)-Norverapamil hydrochloride, LU 48856, (S)-(-)-Nor Verapamil Hydrochloride
InChI formel InChI=1S/C26H36N2O4.ClH/c1-19(2)26(18-27,21-9-11-23(30-4)25(17-21)32-6)13-7-14-28-15-12-20-8-10-22(29-3)24(16-20)31-5;/h8-11,16-17,19,28H,7,12-15H2,1-6H3;1H/t26-;/m0./s1
LEDER Cl.COc1ccc(CCNCCC[C@](C#N)(C(C)C)c2ccc(OC)c(OC)c2)cc1OC
IUPAC-namn (2S)-2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethylamino]-2-propan-2-ylpentanenitrile;hydrochloride
Molekylvikt (g/mol) 477.04
Formel vikt 476.2442