Läs mer
S-(+)-O-Desmethyl Venlafaxine, TRC
Produktkod. 30365170
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365170 50 mg
30352890 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30365170 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29651050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142761-12-4
Molekylformel C16 H25 N O2
Molekylvikt (g/mol) 263.38
Synonym (S)-O-Desmethylvenlafaxine, Phenol, 4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, Phenol, 4-[2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]-, (S)-, 4-[(1S)-2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, (+)-O-Desmethylvenlafaxine, (S)-(+)-O-Desmethylvenlafaxine, S-(+)-O-Desmethyl venlafaxine
IUPAC-namn 4-[(1S)-2-(dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol
LEDER CN(C)C[C@H](c1ccc(O)cc1)C2(O)CCCCC2
Specifikationer
Kemiskt namn eller material S-(+)-O-Desmethyl Venlafaxine
Kvantitet 50 mg
InChI formel InChI=1S/C16H25NO2/c1-17(2)12-15(13-6-8-14(18)9-7-13)16(19)10-4-3-5-11-16/h6-9,15,18-19H,3-5,10-12H2,1-2H3/t15-/m1/s1
Formel vikt 263.1885