Läs mer
(S)-Patulin Methyl Ether, TRC
Produktkod. 30413305
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30413305 5 mg
1 options
Produktkod. 30413305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2065105MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (S)-Patulin Methyl Ether
CAS 123251-09-2
Molekylformel C8H8O4
Kvantitet 5 mg
Synonym (S)-4-Methoxy-4H-furo[3,2-c]pyran-2(6H)-one, (S)-O-Methyl Patulin
InChI formel InChI=1S/C8H8O4/c1-10-8-5-4-7(9)12-6(5)2-3-11-8/h2,4,8H,3H2,1H3/t8-/m0/s1
LEDER CO[C@@H]1C2=CC(=O)OC2=CCO1
Molekylvikt (g/mol) 168.15
Formel vikt 168.0423