Läs mer
S-Pent-4-yn-1-yl Ethanethioate, TRC
Produktkod. 30427065
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427065 250 mg
30410601 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30427065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP283630250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material S-Pent-4-yn-1-yl Ethanethioate
CAS 524689-75-6
Molekylformel C7H10OS
Kvantitet 250 mg
InChI formel InChI=1S/C7H10OS/c1-3-4-5-6-9-7(2)8/h1H,4-6H2,2H3
LEDER CC(=O)SCCCC#C
IUPAC-namn S-pent-4-ynyl ethanethioate
Molekylvikt (g/mol) 142.22
Formel vikt 142.0452