Läs mer
Tocris Bioscience™ (S)-PFI 2 hydrochloride
Negative control of (R)-PFI 2 hydrochloride (Cat. No. 4892)
Brand: Tocris Bioscience™ 5400/10
Beskrivning
(S)-PFI 2 hydrochloride is a negative control of (R)-PFI 2 hydrochloride . Exhibits 500-fold lower activity in a SETD7 enzymatic assay (IC50 = 1 μM) compared to the active enantiomer.Active Analog also available.
Specifikationer
| 97% | |
| 1627607-88-8 | |
| 10 mg |
| 8-Fluoro-1,2,3,4-tetrahydro-N-[(1 S)-2-oxo-2-(1-pyrrolidinyl)-1-[[3-(trifluoromethyl)phenyl]methyl]ethyl]-6-isoquinolinesulfonamide hydrochloride | |
| -20°C |