Läs mer
S-Phenyl Benzenethiosulfonate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | S-Phenyl Benzenethiosulfonate |
| CAS | 1212-08-4 |
| Molekylformel | C12H10O2S2 |
| Kvantitet | 5 g |
| Synonym | NSC 74657, Phenyl Benzenethiosulfonate, Phenyl Phenylthio Sulfone, S-Phenyl Benzenemonothiosulfonate, S-Phenyl Benzenethiosulfonate |
| InChI formel | InChI=1S/C12H10O2S2/c13-16(14,12-9-5-2-6-10-12)15-11-7-3-1-4-8-11/h1-10H |
| LEDER | O=S(=O)(Sc1ccccc1)c2ccccc2 |
| IUPAC-namn | benzenesulfonylsulfanylbenzene |
| Molekylvikt (g/mol) | 250.34 |
| Formel vikt | 250.0122 |