missing translation for 'onlineSavingsMsg'
Learn More

S-Phenyl thioacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7052655
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
934-87-2
C8H8OS
152.211
MFCD00008752
WBISVCLTLBMTDS-UHFFFAOYSA-N
s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl
13630
S-phenyl ethanethioate
CC(=O)SC1=CC=CC=C1
This item is not returnable. View return policy
934-87-2
C8H8OS
152.211
MFCD00008752
WBISVCLTLBMTDS-UHFFFAOYSA-N
s-phenyl thioacetate, thiophenyl acetate, ethanethioic acid, s-phenyl ester, acetic acid, thio-, s-phenyl ester, phenyl thioacetate, thioacetic acid s-phenyl ester, thiacetic acid, s-phenyl ester, pubchem15324, 1-phenylsulfanylethanone, thioacetic acid s-phenyl
13630
S-phenyl ethanethioate
CC(=O)SC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 1.124 g/mL
Boiling Point 99°C to 100°C (6 mmHg)
Flash Point 79°C (174°F)
Linear Formula CH3COSC6H5
Refractive Index 1.57
Quantity 5 g
Formula Weight 152.21
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material S-Phenyl thioacetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'