Läs mer
S-Phenylmercapturic Acid, TRC
Produktkod. 17983525
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17983525 10 mg
17934385 100 mg
17951305 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17983525 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP33560010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4775-80-8
Molekylformel C11 H13 N O3 S
Molekylvikt (g/mol) 239.29
Synonym Alanine, N-acetyl-3-(phenylthio)- (6CI,7CI,8CI), N-Acetyl-S-phenyl-L-cysteine, (2R)-2-Acetamido-3-(phenylsulfanyl)propanoic acid, (R)-2-Acetamido-3-(phenylthio)propanoic acid, NSC 17197, Phenylmercapturic acid, S-Phenylmercapturic acid, L-Cysteine, N-acetyl-S-phenyl-
IUPAC-namn (2R)-2-acetamido-3-phenylsulfanylpropanoic acid
LEDER CC(=O)N[C@@H](CSc1ccccc1)C(=O)O
Specifikationer
Kemiskt namn eller material S-Phenylmercapturic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C11H13NO3S/c1-8(13)12-10(11(14)15)7-16-9-5-3-2-4-6-9/h2-6,10H,7H2,1H3,(H,12,13)(H,14,15)/t10-/m0/s1
Formel vikt 239.0616
Procent renhet >95%