Läs mer
(S)-Pregabalin Methyl Ester Hydrochloride Salt, TRC
Produktkod. 30413862
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30413862 25 mg
1 options
Produktkod. 30413862 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP70481025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Pregabalin Methyl Ester Hydrochloride
CAS 714230-22-5
Molekylformel C9 H19 N O2 . Cl H
Kvantitet 25 mg
Synonym (S)-Pregabalin Methyl Ester Hydrochloride, Hexanoic acid','3-(aminomethyl)-5-methyl-','methyl ester','hydrochloride (1:1)','(3S)-, Hexanoic acid','3-(aminomethyl)-5-methyl-','methyl ester','hydrochloride','(3S)- (9CI)
InChI formel InChI=1S/C9H19NO2.ClH/c1-7(2)4-8(6-10)5-9(11)12-3;/h7-8H,4-6,10H2,1-3H3;1H/t8-;/m0./s1
LEDER Cl.COC(=O)C[C@@H](CN)CC(C)C
IUPAC-namn methyl (3S)-3-(aminomethyl)-5-methylhexanoate;hydrochloride
Molekylvikt (g/mol) 209.71
Formel vikt 209.1183