missing translation for 'onlineSavingsMsg'
Läs mer

(S)-Propranolol-d7 Hydrochloride, TRC

Produktkod. 30417714
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30417714 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Reglering: Begränsningar kan gälla för köp av denna produkt och vi kan behöva begära ytterligare dokumentation från dig.
Produktkod. 30417714 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP8317975MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material (S)-Propranolol-d7 Hydrochloride
CAS 1346617-12-6
Molekylformel C16 H14 D7 N O2 . HCl
Kvantitet 5 mg
Synonym (2S)-1-[(1-Methylethyl-d7)amino]-3-(1-naphthalenyloxy)-2-propanol Hydrochloride, S-(-)-1-(Isopropyl-d7)amino-3-(1-naphthoxy)-2-propanol Hydrochloride, (-)-Propranolol-d7 Hydrochloride, (S)-(-)-Propranolol-d7 Hydrochloride, L-(-)-Propranolol-d7 Hydrochloride, l-Propranolol-d7 Chlorhydrate, l-Propranolol-d7 Hydrochloride
InChI formel InChI=1S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18H,10-11H2,1-2H3;1H/t14-;/m0./s1/i1D3,2D3,12D;
LEDER O[C@H](COC1=CC=CC2=C1C=CC=C2)CNC(C([2H])([2H])[2H])(C([2H])([2H])[2H])[2H].Cl
IUPAC-namn (S)-1-(naphthalen-1-yloxy)-3-((propan-2-yl-d7)amino)propan-2-ol hydrochloride
Molekylvikt (g/mol) 302.85
Formel vikt 302.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.