Läs mer
(S)-Propyl Pramipexole, TRC
Produktkod. 30137466
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137466 50 mg
30138323 100 mg
30004480 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29369050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1147937-31-2
Molekylformel C13 H23 N3 S
Molekylvikt (g/mol) 253.41
Synonym (S)-N6,N6-Dipropyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine, (6S)-4,5,6,7-Tetrahydro-N6,N6-dipropyl-2,6-benzothiazolediamine
IUPAC-namn (6S)-N6,N6-dipropyl-4,5,6,7-tetrahydro-1,3-benzothiazole-2,6-diamine
LEDER CCCN(CCC)[C@H]1CCc2nc(N)sc2C1
Specifikationer
Kemiskt namn eller material (S)-Propyl Pramipexole
Kvantitet 50 mg
InChI formel InChI=1S/C13H23N3S/c1-3-7-16(8-4-2)10-5-6-11-12(9-10)17-13(14)15-11/h10H,3-9H2,1-2H3,(H2,14,15)/t10-/m0/s1
Formel vikt 253.1613