Läs mer
(S)-Repaglinide Ethyl Ester (Repaglinide Impurity), TRC
Produktkod. 30414303
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30414303 10 mg
1 options
Produktkod. 30414303 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR14450510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ethyl 2-Ethoxy-4-[2-(((1S)-3-methyl-1-[2-(piperidin-1-yl)phenyl]butyl]amino)-2-oxoethyl)-benzoate (Repaglinide Ethyl Ester)
CAS 147770-06-7
Molekylformel C29 H40 N2 O4
Kvantitet 10 mg
Synonym 2-Ethoxy-4-[2-[[(1S)-3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]-benzoic acid ethyl ester, Benzoic acid','2-ethoxy-4-[2-[[3-methyl-1-[2-(1-piperidinyl)phenyl]butyl]amino]-2-oxoethyl]-','ethyl ester','(S)-, (S)-2-Ethoxy-4-[[[N-[1-(2-piperidinophenyl)-3-methylbutyl]amino]carbonyl]methyl]benzoic acid ethyl ester, (S)-Ethyl 2-ethoxy-4-[[[N-[1-(2-piperidinophenyl)-3-methyl-1-butyl]amino]carbonyl]methyl]benzoate
InChI formel InChI=1S/C29H40N2O4/c1-5-34-27-19-22(14-15-24(27)29(33)35-6-2)20-28(32)30-25(18-21(3)4)23-12-8-9-13-26(23)31-16-10-7-11-17-31/h8-9,12-15,19,21,25H,5-7,10-11,16-18,20H2,1-4H3,(H,30,32)/t25-/m0/s1
LEDER CCOC(=O)c1ccc(CC(=O)N[C@@H](CC(C)C)c2ccccc2N3CCCCC3)cc1OCC
IUPAC-namn ethyl 2-ethoxy-4-[2-[[(1S)-3-methyl-1-(2-piperidin-1-ylphenyl)butyl]amino]-2-oxoethyl]benzoate
Molekylvikt (g/mol) 480.64
Formel vikt 480.2988