Läs mer
(S)-Ropivacaine, TRC
Produktkod. 30420439
Change view
Kvantitet:
200 mg
Produktkod. Kvantitet
30420439 200 mg
1 options
Produktkod. 30420439 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR674995200MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Ropivacaine
CAS 84057-95-4
Molekylformel C17 H26 N2 O
Kvantitet 200 mg
Synonym 2-Piperidinecarboxamide','N-(2,6-dimethylphenyl)-1-propyl-','(2S)-, 2-Piperidinecarboxamide','N-(2,6-dimethylphenyl)-1-propyl-','(S)-, (2S)-N-(2,6-Dimethylphenyl)-1-propyl-2-piperidinecarboxamide, (-)-LEA 103, (-)-Ropivacaine, (2S)-N-(2,6-Dimethylphenyl)-1-propyl-2-piperidinecarboxamide, (2S)-N-(2,6-Dimethylphenyl)-1-propylpiperidine-2-carboxamide, (S)-(-)-Ropivacaine, Anapeine, LEA 103, Narop, Naropeine, Naropine, Ropivacaine, S-Ropivacaine
InChI formel InChI=1S/C17H26N2O/c1-4-11-19-12-6-5-10-15(19)17(20)18-16-13(2)8-7-9-14(16)3/h7-9,15H,4-6,10-12H2,1-3H3,(H,18,20)/t15-/m0/s1
LEDER CCCN1CCCC[C@H]1C(=O)Nc2c(C)cccc2C
IUPAC-namn (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide
Molekylvikt (g/mol) 274.4
Formel vikt 274.2045