Läs mer
(S)-Ruxolitinib Phosphate, TRC
Produktkod. 30180285
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180285 10 mg
30179923 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180285 Leverantör Toronto Research Chemicals Leverantörsnummer TRCR71311510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2703484-89-1
Molekylformel C17H18N6 • H3PO4
Molekylvikt (g/mol) 404.37
IUPAC-namn (S)-3-(4-(1H-pyrrolo[2,3-d]pyrimidin-4-yl)-1H-pyrazol-1-yl)-3-cyclopentylpropanenitrile phosphate
LEDER O=P(O)(O)O.N#CC[C@@H](C1CCCC1)N2C=C(C=N2)C3=C4C=CN=C4NC=N3
Specifikationer
Kemiskt namn eller material (S)-Ruxolitinib Phosphate
Kvantitet 10 mg
InChI formel InChI=1S/C17H18N6.H3O4P/c18-7-5-15(12-3-1-2-4-12)23-10-13(9-22-23)16-14-6-8-19-17(14)21-11-20-16;1-5(2,3)4/h6,8-12,15H,1-5H2,(H,19,20,21);(H3,1,2,3,4)/t15-;/m0./s1
Formel vikt 404.14