missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-1,2-Bis(4-nitrophenyl)-1,2-ethanediamine dihydrochloride, 95%, ee 99%, Thermo Scientific™

Catalog Number p-7028579
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1052707-07-9
C14H16Cl2N4O4
375.206
MFCD09265322
PDPYGNJVCKPVGM-AXEKQOJOSA-N
1s, 2s-1,2-bis 4-nitrophenyl ethylenediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine dihydrochloride, 1r,2r-1,2-bis 4-nitrophenyl ethane-1,2-diaminedihydrochloride, 1s, 2s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, ee
45925681
(1S,2S)-1,2-bis(4-nitrophenyl)ethane-1,2-diamine;dihydrochloride
C1=CC(=CC=C1C(C(C2=CC=C(C=C2)[N+](=O)[O-])N)N)[N+](=O)[O-].Cl.Cl
This item is not returnable. View return policy
1052707-07-9
C14H16Cl2N4O4
375.206
MFCD09265322
PDPYGNJVCKPVGM-AXEKQOJOSA-N
1s, 2s-1,2-bis 4-nitrophenyl ethylenediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine dihydrochloride, 1r,2r-1,2-bis 4-nitrophenyl ethane-1,2-diaminedihydrochloride, 1s, 2s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, 1s,2s-1,2-bis 4-nitrophenyl ethane-1,2-diamine-hydrogen chloride 1/2, s,s-1,2-bis 4-nitrophenyl-1,2-ethanediamine dihydrochloride, ee
45925681
(1S,2S)-1,2-bis(4-nitrophenyl)ethane-1,2-diamine;dihydrochloride
C1=CC(=CC=C1C(C(C2=CC=C(C=C2)[N+](=O)[O-])N)N)[N+](=O)[O-].Cl.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 196°C
Quantity 250 mg
Optical Rotation −83° (c=1 in Water)
Formula Weight 375.23
Percent Purity 95%
Assay ee 99%
Chemical Name or Material (S,S)-1,2-Bis(4-nitrophenyl)-1,2-ethanediamine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'