missing translation for 'onlineSavingsMsg'
Läs mer

(S,S)-2,8-Diazabicyclo[4.3.0]nonane, TRC

Produktkod. 30355842
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355842 10 g
30363261 100 mg
30359424 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30355842 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD41687110G

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151213-40-0
Molekylformel C7 H14 N2
Molekylvikt (g/mol) 126.2
Synonym 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS,7aS)-, 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS-cis)-, (1S,6S)-2,8-Diazabicyclo[4.3.0]nonane, (1S,6S)-cis-2,8-Diazabicyclo[4.3.0]nonane, (4AS,7aS)-Octahydro-1H-pyrrole[3,4-b]pyridine, (S,S)-2,8-Diazabicyclo[4.3.0]nonane
IUPAC-namn (4aS,7aS)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine
LEDER C1CN[C@@H]2CNC[C@@H]2C1

Specifications

Kemiskt namn eller material (S,S)-2,8-Diazabicyclo[4.3.0]nonane
Kvantitet 10 g
InChI formel InChI=1S/C7H14N2/c1-2-6-4-8-5-7(6)9-3-1/h6-9H,1-5H2/t6-,7+/m0/s1
Formel vikt 126.1157
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.