Läs mer
(S,S)-2,8-Diazabicyclo[4.3.0]nonane, TRC
Produktkod. 30363261
Change view
Kvantitet:
100 mg
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363261 100 mg
30355842 10 g
30359424 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30363261 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD416871100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 151213-40-0
Molekylformel C7 H14 N2
Molekylvikt (g/mol) 126.2
Synonym 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS,7aS)-, 1H-Pyrrolo[3,4-b]pyridine, octahydro-, (4aS-cis)-, (1S,6S)-2,8-Diazabicyclo[4.3.0]nonane, (1S,6S)-cis-2,8-Diazabicyclo[4.3.0]nonane, (4AS,7aS)-Octahydro-1H-pyrrole[3,4-b]pyridine, (S,S)-2,8-Diazabicyclo[4.3.0]nonane
IUPAC-namn (4aS,7aS)-2,3,4,4a,5,6,7,7a-octahydro-1H-pyrrolo[3,4-b]pyridine
LEDER C1CN[C@@H]2CNC[C@@H]2C1
Specifikationer
Kemiskt namn eller material (S,S)-2,8-Diazabicyclo[4.3.0]nonane
Kvantitet 100 mg
InChI formel InChI=1S/C7H14N2/c1-2-6-4-8-5-7(6)9-3-1/h6-9H,1-5H2/t6-,7+/m0/s1
Formel vikt 126.1157