Läs mer
(S,S)-Hydroxy Bupropion, TRC
Produktkod. 30401799
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30401799 1 mg
30394507 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30401799 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH8306701MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 192374-14-4
Molekylformel C13 H18 Cl N O2 . Cl H
Molekylvikt (g/mol) 292.2
Synonym (2S,3S)-2-(3-Chlorophenyl)-3,5,5-trimethyl-2-morpholinol, Radafaxine
IUPAC-namn (2S,3S)-2-(3-chlorophenyl)-3,5,5-trimethylmorpholin-2-ol;hydrochloride
LEDER Cl.C[C@@H]1NC(C)(C)CO[C@@]1(O)c2cccc(Cl)c2
Specifikationer
Kemiskt namn eller material (S,S)-hydroxy Bupropion
Kvantitet 1 mg
InChI formel InChI=1S/C13H18ClNO2.ClH/c1-9-13(16,17-8-12(2,3)15-9)10-5-4-6-11(14)7-10;/h4-7,9,15-16H,8H2,1-3H3;1H/t9-,13+;/m0./s1
Formel vikt 291.0793