Läs mer
(S,S)-Oxaliplatin, TRC
Produktkod. 30417479
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30417479 10 mg
1 options
Produktkod. 30417479 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO84508010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material cis-[(1S,2S)-1,2-cyclohexanediamine-N,N'][oxalato(2-)-O,O']platinum
CAS 61758-77-8
Molekylformel C8 H14 N2 O4 Pt
Kvantitet 10 mg
Synonym Platinum','[(1S,2S)-1,2-cyclohexanediamine-kappaN1,kappaN2][ethanedioato(2-)-kappaO1,kappaO2]-','(SP-4-2)-, Platinum','(1,2-cyclohexanediamine-kappaN,kappaN')[ethanedioato(2-)-kappaO1,kappaO2]-','[SP-4-2-[1S-(trans)]]-, Platinum','[(1S,2S)-1,2-cyclohexanediamine-kappaN,kappaN'][ethanedioato(2-)-kappaO1,kappaO2]- (9CI), 1,2-Cyclohexanediamine','platinum complex','(1S-trans)-, [(1S,2S)-1,2-Cyclohexanediamine-kappaN1,kappaN2][ethanedioato(2-)-kappaO1,kappaO2]platinum, NSC 266042, Platinum','(1,2-cyclohexanediamine-N,N')[ethanedioato(2-)-O,O']-','[SP-4-2-(1S-trans)]-, (SP-4-2)-[(1S,2S)-Cyclohexane-1,2-diamine-kappaN,kappaN′][ethanedioato(2-)-kappaO1,kappaO2]platinum, Oxaliplatin Imp. D (EP), (S,S)-Oxaliplatin, Oxaliplatin Related Compound D (USP)
InChI formel InChI=1S/C6H14N2.C2H2O4.Pt/c7-5-3-1-2-4-6(5)8;3-1(4)2(5)6;/h5-6H,1-4,7-8H2;(H,3,4)(H,5,6);/q;;+2/p-2/t5-,6-;;/m0../s1
LEDER O=C1C(=O)[O-][Pt+2]2([NH2][C@H]3CCCC[C@@H]3[NH2]2)[O-]1
Molekylvikt (g/mol) 397.29
Formel vikt 397.0601