Läs mer
(S,S,S)-Enalapril Maleate, TRC
Produktkod. 30040545
Change view
Kvantitet:
100 mg
250 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040545 100 mg
30010476 250 mg
30006529 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE555250100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 76095-16-4
Synonym Enalapril Maleate∣L-Proline, N-[(1S)-1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl-, (2Z)-2-butenedioate (1:1)∣L-Proline, 1-[N-[1-(ethoxycarbonyl)-3-phenylpropyl]-L-alanyl]-, (S)-, (Z)-2-butenedioate (1:1)∣Acapril∣Acetensil∣Alphrin∣Amprace∣Analept∣Antiprex∣BQL∣Benalipril∣Biocronil∣Bitensil∣Cardiovet∣Controlvas∣Converten∣Convertin∣ENAP∣Ednyt∣Elfonal∣Enacard∣Enace∣Enaladil∣Enalapril LPH∣Enalapril maleate∣Enaloc∣Enapren∣Enapril∣Enaprin∣Enaril∣Envas∣Ezapril∣Glioten∣Hipoartel∣Hytrol∣Innovace∣Innovade∣Inoprilat∣Invoril∣Kenopril∣Lapril∣Lotrial∣MK 421∣MK 421 maleate∣Minipril∣Naprilene∣Naritec∣Nuril∣Olivin∣Pres∣Presil∣Pressotec∣Renavace∣Renitec∣Renitek∣Reniten∣Renivace∣Repantril∣Sintec∣Tenace∣Unaril∣Unipril∣Vasopril∣Vasotec∣Xanef
Specifikationer
Formatera Neat
Molekylformel C24H32N2O9
Molekylvikt (g/mol) 492.52, 492.53
InChI-nyckel OYFJQPXVCSSHAI-BMSLDGIRSA-N
LEDER OC(=O)\C=C/C(O)=O.CCOC(=O)[C@@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O
IUPAC-namn (2S)-1-[(2S)-2-{[(2R)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid; (2Z)-but-2-enedioic acid
Produkttyp Metabolite∣API