Läs mer
S-(-)-Tolterodine D-Tartrate, TRC
Produktkod. 30455971
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30455971 5 mg
30455429 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30455971 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT5357965MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 124937-54-8
Molekylformel C22 H31 N O . C4 H6 O6
Molekylvikt (g/mol) 475.57
Synonym 2-[(1S)-3-[Bis(1-methylethyl)amino]-1-phenylpropyl]-4-methyl-phenol D-Tartrate, (S)-Tolterodine D-Tartrate
IUPAC-namn (2S,3S)-2,3-dihydroxybutanedioic acid;2-[(1S)-3-(diisopropylamino)-1-phenyl-propyl]-4-methyl-phenol
LEDER CC(C)N(CC[C@@H](c1ccccc1)c2cc(C)ccc2O)C(C)C.O[C@@H]([C@H](O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material S-(-)-Tolterodine D-tartrate
Kvantitet 5 mg
InChI formel InChI=1S/C22H31NO.C4H6O6/c1-16(2)23(17(3)4)14-13-20(19-9-7-6-8-10-19)21-15-18(5)11-12-22(21)24;5-1(3(7)8)2(6)4(9)10/h6-12,15-17,20,24H,13-14H2,1-5H3;1-2,5-6H,(H,7,8)(H,9,10)/t20-;1-,2-/m00/s1
Formel vikt 475.257