Läs mer
(S)-Vigabatrin, TRC
Produktkod. 30456475
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30456475 25 mg
30456761 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30456475 Leverantör Toronto Research Chemicals Leverantörsnummer TRCV25300025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 74046-07-4
Molekylformel C6 H11 N O2
Molekylvikt (g/mol) 129.16
Synonym S-Vigabatrin
IUPAC-namn (4S)-4-aminohex-5-enoic acid
LEDER N[C@@H](CCC(=O)O)C=C
Specifikationer
Kemiskt namn eller material (S)-Vigabatrin
Kvantitet 25 mg
InChI formel InChI=1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/t5-/m1/s1
Formel vikt 129.079