missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ S100A4 Ab-8 Rabbit Polyclonal Antibody with BSA and azide
Epredia™ S100 A4 Ab-8, Rabbit Polyclonal Antibody provides accurate, reproducible results.
1215.00 SEK - 7298.10 SEK
Specifications
Antigen | S100 A4 Ab-8 |
---|---|
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
12673147
|
Epredia™
RB-1804-A0 |
Concentrated | 100 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12683147
|
Epredia™
RB-1804-A1 |
Concentrated | 500 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12663147
|
Epredia™
RB-1804-A |
Concentrated | 1 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12613157
|
Epredia™
RB-1804-R7 |
Prediluted | 7 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
The S100A4 protein belongs to the S100 subfamily, now designated S100A1/S100A13. This antibody does not cross react with other S100 proteins. Anti-S100A4 labels fibroblasts, neuronal structures, T-lymphocytes, plasma cells and Kupffer cells in normal tissues.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, Rat, Cow, Horse, Cat and Dog. Others not known.
Epitope: Not determined
Immunogen: Purified recombinant human S100 A4 monomer expressed in E. coli
Molecular Weight: 10-12kDa
Positive Control: Tonsil
Cellular Localization: Cytoplasmic, membrane and nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
S100 A4 Ab-8 | |
Polyclonal | |
Rabbit | |
Cancer and Tumor Biology | |
Purified recombinant human S100 A4 monomer expressed in E. coli | |
Primary |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Bovine, Canine, Equine, Feline, Human, Mouse | |
IgG |