missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100 beta Mouse anti-Human, Porcine, Rat, Clone: 1G10F4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66616-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
S100B is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and are involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100B may function in neurite extension, proliferation of melanoma cells, stimulation of Ca2+ fluxes, inhibition of PKC-mediated phosphorylation, astrocytosis and axonal proliferation, and inhibition of microtubule assembly. Chromosomal rearrangements and altered expression of this gene have been implicated in several diseases, including Alzheimer′s disease, Down′s syndrome, epilepsy, amyotrophic lateral sclerosis, melanoma, and type I diabetes.Specifications
S100 beta | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P04271, P04631 | |
S100B | |
S100 beta Fusion Protein Ag7440 | |
20 μL | |
Primary | |
Porcine, Rat, Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1G10F4 | |
Unconjugated | |
S100B | |
NEF, Protein S100 B, S 100 protein beta chain, S 100 protein subunit beta, S100, S100 calcium binding protein B, S100B, S100beta | |
Mouse | |
Protein G | |
RUO | |
100623510, 25742, 6285 | |
-20°C | |
Liquid |