missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ S100 Monoclonal Antibody (6G1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA120103
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA1-20103 detects S100 in human samples.
S100 belongs to the family of calcium binding proteins such as calmodulin and troponin C. S100A is composed of an alpha and beta chain whereas S100B is composed of two beta chains. S100 protein is also expressed in the antigen presenting cells such as the Langerhans cells in skin and interdigitating reticulum cells in the paracortex of lymph nodes.
Specifikationer
| S100 | |
| Monoclonal | |
| 4 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| P04271, P23297 | |
| S100A1, S100B | |
| Human S-100 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Affinity Purification, ELISA, Western Blot | |
| 6G1 | |
| Unconjugated | |
| S100B | |
| AI266795; AI850290; Bpb; hypothetical protein LOC525716; NEF; Protein S100-A1; protein S100-B; S 100; S100; S100 alpha; S100 calcium binding protein A1; S100 calcium binding protein B; S100 calcium-binding protein A1; S100 calcium-binding protein B; S100 calcium-binding protein beta (neural); S100 calcium-binding protein, beta (neural); S-100 calcium-binding protein, beta chain; S-100 protein alpha chain; S-100 protein beta chain; S-100 protein subunit alpha; S-100 protein subunit beta; S100 protein, alpha polypeptide; S100 protein, beta polypeptide; S100 protein, beta polypeptide, neural; S100a; S100A1; S100alpha; S100-alpha; S100B; S100-B; S100beta; S100P | |
| Mouse | |
| Protein A | |
| RUO | |
| 6271, 6285 | |
| 4°C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering