missing translation for 'onlineSavingsMsg'
Learn More

S100A8 Recombinant Human Protein, His Tag

S100A8 Recombinant Human Protein, His Tag

Brand:  Invitrogen 11138H08B25

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15258275

  • 1640.00 SEK / 25µg


Estimated Shipment date
from Supplier 04-02-2019
Add to basket

Description

Description

S100 Calcium Binding Protein A8 (S100A8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in performing cell-based assays and protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

This recombinant protein was expressed from a DNA sequence encoding the human S100A8 (NP_002955.2) (Leu 2-Glu 93) fused to a polyhistidine tag at the N-terminus.

Activity:
1. Measured by its ability to bind recombinant human S100A9 in a functional ELISA.
2.Measured by its ability to inhibit the proliferation of MCF7 human breast adenocarcinoma cells. The ED50 for this effect is typically 3-15μg/ml.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.

This protein is manufactured by SINO Biological.
Specifications

Specifications

Protein assays and analysis
Lyophilized
S100A8 Recombinant Human Protein
≥90% by SDS-PAGE
–20°C
5 Pack(s)
60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8
5 Vials of lyophilized protein
Assay standard
Insect
Human
Lyophilized
12.2kDa
8 ±0.5
5 x 5μg
For research use only. Not for use in diagnostic procedures
Other proteins
S100A8 Recombinant Human Protein
S100A8
Other proteins
S100A8 Recombinant Human Protein
His Tag
Videos