Läs mer
S100A9 Mouse anti-Human, Clone: MRP14/840, Abnova™
Mouse monoclonal antibody raised against full length recombinant human S100A9.
Brand: Abnova MAB13429.100ug
Beskrivning
The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein may function in the inhibition of casein kinase and altered expression of this protein is associated with the disease cystic fibrosis. [provided by RefSeq]
Specifikationer
| S100A9 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| 60B8AG/CAGB/CFAG/CGLB/L1AG/LIAG/MAC387/MIF/MRP14/NIF/P14 | |
| Mouse | |
| PEG precipitation | |
| RUO | |
| 6280 | |
| Store at 4°C. | |
| IgM κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| MRP14/840 | |
| Mouse monoclonal antibody raised against full length recombinant human S100A9. | |
| In 1mM PBS (0.05% BSA, 0.05% sodium azide). | |
| S100A9 | |
| Recombinant protein corresponding to full length human S100A9. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.