missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sa Cas9 Rabbit anti-Bacteria, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Sa Cas9 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16885255
|
Proteintech
26927-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16875255
|
Proteintech
26927-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects the Cas9 of Staphylococcus aureus (SaCas9). It does not detect the SpCas9.
Cas9 (CRISPR associated protein 9) is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in Streptococcus pyogenes (Sp).Specifications
Sa Cas9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Bacteria | |
Cas9, Sa Cas9, SaCas9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Sa Cas9 Fusion Protein Ag25669 | |
Primary | |
-20°C |