Läs mer
Sabinene Hydrate, TRC
Produktkod. 30428260
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30428260 100 mg
30424422 1 g
30426981 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428260 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS807585100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 546-79-2
Molekylformel C10 H18 O
Molekylvikt (g/mol) 154.25
Synonym Bicyclo[3.1.0]hexan-2-ol, 2-methyl-5-(1-methylethyl)-, 4-Thujanol (6CI,7CI,8CI), 2-Methyl-5-(1-methylethyl)bicyclo[3.1.0]hexan-2-ol, 5-Isopropyl-2-methylbicyclo[3.1.0]hexan-2-ol, Sabinene hydrate, 4-Thujanol
IUPAC-namn 4-methyl-1-propan-2-ylbicyclo[3.1.0]hexan-4-ol
LEDER CC(C)C12CCC(C)(O)C1C2
Specifikationer
Kemiskt namn eller material 4-Thujanol
Kvantitet 100 mg
InChI formel InChI=1S/C10H18O/c1-7(2)10-5-4-9(3,11)8(10)6-10/h7-8,11H,4-6H2,1-3H3
Formel vikt 154.1358