Läs mer
Saccharin Sodium Dihydrate, 98.0+%, Extra Pure, SLR, Fisher Chemical
Handla allt Fisher Chemical ProdukterSpecifikationer
Specifikationer
| CAS | 6155-57-3 |
| Färg | White |
| Fysisk form | Solid |
| Kvantitet | 100 g |
| Molekylformel | C7H9NNaO5S |
| MDL-nummer | 151213 |
| Synonym | saccharin sodium dihydrate, saccharin sodium, sodium saccharin, saccharin soluble, saccharin sodium salt hydrate, 1,2-benzisothiazol-3 2h-one, 1,1-dioxide, sodium salt, dihydrate, o-sulfobenzimide sodium salt, 2-sulfobenzoic acid imide sodium salt, saccharin sodium salt dihydrate, c7h4nnao3s.2h2o |
| InChI-nyckel | UQKRDMFNUSMUCB-UHFFFAOYSA-N |
| LEDER | C1=CC=C2C(=C1)C(=O)NS2(=O)=O.O.O.[Na] |
| IUPAC-namn | 1,1-dioxo-1,2-benzothiazol-3-one;sodium;dihydrate |
| Visa mer |